Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163467.1 | NP_001156939.1 | 4 | 281 | ||
NM_001163468.1 | NP_001156940.1 | 4 | 687 | |||
NM_001163469.1 | NP_001156941.1 | 4 | 687 | |||
NM_013629.2 | NP_038657.2 | 4 | 687 | |||
GenBank mRNA | AJ133721.1 | - | 4 | 281 | ||
AJ242864.1 | - | 4 | 687 | |||
AK019146.1 | - | 4 | 591 | |||
AK088714.1 | - | 4 | 578 | |||
BC021147.1 | - | 4 | 559 | |||
BC050825.1 | - | 4 | 499 | 6518647 | ||
BC057340.1 | - | 4 | 573 | 6827816 | ||
BC060177.1 | - | 4 | 573 | 6827816 |