Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Greb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252071.1 | NP_001239000.1 | 7, 8 | 1220 | ||
NM_015764.4 | NP_056579.2 | 9, 10 | 1542 | |||
GenBank mRNA | AF180470.1 | - | 8, 9 | 1198 | ||
AK147337.1 | - | 7, 8 | 1070 | |||
AK157808.1 | - | 7, 8 | 1154 | |||
BC160931.1 | - | 8, 9 | 1152 |