Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Greb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252071.1 | NP_001239000.1 | 33 | 6813 | ||
NM_015764.4 | NP_056579.2 | 35 | 7219 | |||
GenBank mRNA | AB093248.1 | - | 18 | 4148 | ||
AF180470.1 | - | 34 | 6875 | |||
AK017875.1 | - | 10 | 2436 | |||
AK033573.1 | - | 5 | 1379 | |||
AK147337.1 | - | 33 | 6747 | |||
CN533010.1 | - | 1 | 417 |