Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Corin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122756.1 | NP_001116228.1 | 7 | 1039 | ||
NM_016869.3 | NP_058565.2 | 7 | 1196 | |||
GenBank mRNA | AB013874.1 | - | 7 | 1196 | ||
BC093485.1 | - | 7 | 972 | 30648458 | ||
BC138339.1 | - | 7 | 1179 | 8861359 | ||
BC138340.1 | - | 7 | 1179 | 8861360 | ||
BC145243.1 | - | 7 | 1179 | 9053795 |