Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019580.4 | NP_062526.1 | 4 | 681 | ||
GenBank mRNA | AF212860.1 | - | 4 | 662 | ||
AK004583.1 | - | 4 | 694 | |||
AK005361.1 | - | 4 | 682 | |||
AK150807.1 | - | 4 | 703 | |||
BC003902.1 | - | 4 | 640 | 3601420 |