Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wsb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019653.3 | NP_062627.3 | 4 | 876 | ||
GenBank mRNA | AF033186.1 | - | 4 | 573 | ||
AK019206.2 | - | 4 | 853 | |||
AK145974.1 | - | 4 | 855 | |||
AK146752.1 | - | 4 | 851 | |||
AK150394.1 | - | 4 | 851 | |||
AK150600.1 | - | 4 | 851 | |||
AK152545.1 | - | 4 | 854 | |||
AK159905.1 | - | 4 | 851 | |||
AK167174.1 | - | 4 | 853 | |||
AK167394.1 | - | 4 | 854 | |||
AK167942.1 | - | 4 | 851 | |||
AK168468.1 | - | 4 | 854 | |||
AK168565.1 | - | 4 | 854 | |||
AK169058.1 | - | 4 | 851 | |||
AK200135.1 | - | 2 | 335 | |||
BC053391.1 | - | 4 | 627 | 30073116 |