Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204165.1 | NP_001191094.1 | 11 | 1127 | ||
NM_001204166.1 | NP_001191095.1 | 8 | 1019 | |||
NM_019688.2 | NP_062662.1 | 10 | 1073 | |||
GenBank mRNA | AB021132.1 | - | 11 | 1102 | ||
AB037668.1 | - | 2 | 462 | |||
AB445011.1 | - | 8 | 1011 | |||
AF115480.1 | - | 10 | 1001 | |||
AK004874.1 | - | 2 | 119 | |||
AK220281.1 | - | 11 | 1127 | |||
BC139822.1 | - | 11 | 1059 | 40141811 |