Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019766.4 | NP_062740.1 | 8 | 972 | ||
GenBank mRNA | AB024935.1 | - | 8 | 780 | ||
AK008805.1 | - | 8 | 1003 | |||
AK010636.1 | - | 8 | 967 | |||
AK075932.1 | - | 8 | 839 | |||
AK075987.1 | - | 8 | 883 | |||
AK077538.1 | - | 8 | 973 | |||
AK088321.1 | - | 1 | 54 | |||
AK167443.1 | - | 8 | 946 | |||
AK168073.1 | - | 8 | 882 | |||
AK168210.1 | - | 8 | 882 | |||
AK168721.1 | - | 8 | 822 | |||
AY281130.1 | - | 8 | 999 | |||
BC003708.1 | - | 8 | 863 | 3489418 | ||
BC085264.1 | - | 8 | 869 | 6392001 |