Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159532.1 | NP_001153004.1 | 8 | 1594 | ||
NM_019784.5 | NP_062758.3 | 8 | 1594 | |||
GenBank mRNA | AB000682.1 | - | 8 | 1591 | ||
AK016434.1 | - | 8 | 1626 | |||
AK016448.1 | - | 8 | 1584 | |||
AK016471.1 | - | 8 | 1565 | |||
BC100432.1 | - | 8 | 1594 | 30917866 |