Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204239.1 | NP_001191168.1 | 4, 5 | 773 | ||
NM_001204253.1 | NP_001191182.1 | 3, 4 | 621 | |||
NM_019985.3 | NP_064369.1 | 5, 6 | 869 | |||
GenBank mRNA | AF201457.1 | - | 5, 6 | 580 | ||
AK007925.1 | - | 5, 6 | 849 | |||
AK154961.1 | - | 124 | ||||
BC064054.1 | - | 5, 6 | 869 | 574821 | ||
EF070191.1 | - | 4, 5 | 463 | |||
EF070192.1 | - | 3, 4 | 308 |