Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dolpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290508.1 | NP_001277437.1 | 9 | 1208 | ||
NM_001290509.1 | NP_001277438.1 | 7 | 872 | |||
NM_020329.4 | NP_065062.1 | 8 | 1001 | |||
GenBank mRNA | AB030189.1 | - | 8 | 841 | ||
AK002544.1 | - | 9 | 1047 | |||
AK010797.1 | - | 4 | 438 | |||
AK044141.1 | - | 8 | 840 | |||
AK053923.1 | - | 8 | 830 | |||
AK088065.1 | - | 8 | 846 | |||
BC026544.1 | - | 8 | 825 | 4975127 |