Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042484.1 | NP_001035949.1 | 5 | 492 | ||
NM_020585.2 | NP_065610.2 | 4 | 705 | |||
GenBank mRNA | AB041568.1 | - | 4 | 640 | ||
AK048175.1 | - | 5 | 468 | |||
AK077582.1 | - | 5 | 466 | |||
AK150462.1 | - | 5 | 486 | |||
AK150696.1 | - | 4 | 647 | |||
AK151353.1 | - | 5 | 486 | |||
AK152520.1 | - | 5 | 486 | |||
AK152606.1 | - | 5 | 465 | |||
AK153213.1 | - | 5 | 486 | |||
AK158227.1 | - | 4 | 611 | |||
AK170129.1 | - | 5 | 471 | |||
AK179247.1 | - | 4 | 314 | |||
AK205367.1 | - | 3 | 205 | |||
BC016894.1 | - | 5 | 444 | 4223937 | ||
BC019697.1 | - | 4 | 630 | |||
BC058523.1 | - | 4 | 549 | 5717934 | ||
BC093518.1 | - | 5 | 462 | 3157520 | ||
BC103796.1 | - | 5 | 457 | 6519018 |