Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005471.4 | NP_005462.1 | 2 | 163 | ||
XM_005268348.1 | XP_005268405.1 | 2 | 420 | |||
XM_006714747.2 | XP_006714810.1 | 3 | 187 | |||
XM_011537537.1 | XP_011535839.1 | 3 | 482 | |||
GenBank mRNA | AF029914.1 | - | 1 | 109 | ||
AF048826.1 | - | 1 | 96 | |||
AJ002231.1 | - | 1 | 855 | |||
AK022779.1 | - | 2 | 135 | |||
AK290686.1 | - | 1 | 117 | |||
AK296452.1 | - | 2 | 135 | |||
BC012853.2 | - | 1 | 101 | 3864091 | ||
BC020769.1 | - | 2 | 135 | 4242886 | ||
BC022322.1 | - | 2 | 135 | 4243999 | ||
D31766.1 | - | 3 | 497 | |||
DA661926.1 | - | 2 | 163 |