Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015053.1 | NP_001015053.1 | 6 | 900 | ||
NM_005474.4 | NP_005465.2 | 6 | 897 | |||
XM_005256905.1 | XP_005256962.1 | 6 | 895 | |||
XM_005256906.4 | XP_005256963.1 | 6 | 856 | |||
XM_011524149.1 | XP_011522451.1 | 6 | 871 | |||
XM_011524150.1 | XP_011522452.1 | 5 | 647 | |||
XM_017023988.1 | XP_016879477.1 | 8 | 1009 | |||
XM_017023989.1 | XP_016879478.1 | 8 | 975 | |||
XM_017023990.1 | XP_016879479.1 | 8 | 1000 | |||
XM_017023991.1 | XP_016879480.1 | 8 | 1006 | |||
XM_017023992.1 | XP_016879481.1 | 8 | 1010 | |||
XM_017023993.1 | XP_016879482.1 | 8 | 982 | |||
XM_017023994.1 | XP_016879483.1 | 4 | 963 | |||
XM_017023995.1 | XP_016879484.1 | 6 | 865 | |||
XM_017023996.1 | XP_016879485.1 | 7 | 1072 | |||
GenBank mRNA | AB011172.2 | - | 6 | 869 | ||
AF132608.1 | - | 5 | 565 | |||
AK294760.1 | - | 6 | 865 | |||
BC013140.1 | - | 3 | 347 | |||
BC051824.1 | - | 6 | 776 | 6043491 |