Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021369.2 | NP_067344.2 | 5 | 1522 | ||
GenBank mRNA | AJ245706.1 | - | 5 | 1360 | ||
AK149262.1 | - | 5 | 1463 | |||
AY574265.1 | - | 5 | 1280 | |||
BC031985.1 | - | 5 | 1522 | 4501558 |