Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021369.2 | NP_067344.2 | 6 | 2808 | ||
GenBank mRNA | AK149262.1 | - | 6 | 2749 | ||
BC031985.1 | - | 6 | 2808 | 4501558 |