Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198790.1 | NP_001185719.1 | 7 | 1251 | ||
NM_001198791.1 | NP_001185720.1 | 7 | 1233 | |||
NM_001198792.1 | NP_001185721.1 | 6 | 1192 | |||
NM_021515.3 | NP_067490.1 | 6 | 1210 | |||
GenBank mRNA | AJ010108.1 | - | 6 | 1118 | ||
AJ010109.1 | - | 6 | 1208 | |||
AK046613.1 | - | 7 | 1252 | |||
AK089270.1 | - | 7 | 1232 | |||
BC014802.1 | - | 7 | 1178 | 4505142 | ||
BC054366.1 | - | 6 | 1199 | 5346232 |