Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1aip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160180.1 | NP_001153652.1 | 3 | 2087 | ||
NM_001160181.1 | NP_001153653.1 | 3 | 2035 | |||
NM_001160182.1 | NP_001153654.1 | 3 | 1966 | |||
NM_022329.4 | NP_071724.1 | 2 | 1975 | |||
GenBank mRNA | AK032697.1 | - | 2 | 1919 | ||
AK075980.1 | - | 1 | 1206 | |||
AK150117.1 | - | 3 | 2090 | |||
AK150303.1 | - | 3 | 2004 | |||
AK152137.1 | - | 3 | 2011 | |||
BC053726.1 | - | 3 | 1990 | |||
BC064017.1 | - | 2 | 1940 | 3995710 | ||
BC092059.1 | - | 3 | 1993 | 4019181 |