Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023162.4 | NP_075651.1 | 3 | 465 | ||
GenBank mRNA | AF230339.1 | - | 3 | 338 | ||
AK003714.1 | - | 3 | 466 | |||
AK005976.1 | - | 4 | 438 | |||
AK019152.1 | - | 4 | 442 | |||
BC043016.1 | - | 4 | 402 | 5683859 |