Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023162.4 | NP_075651.1 | 4 | 512 | ||
GenBank mRNA | AF230339.1 | - | 4 | 385 | ||
AK003714.1 | - | 4 | 513 | |||
AK005976.1 | - | 5 | 485 | |||
AK019152.1 | - | 5 | 489 | |||
BC043016.1 | - | 5 | 449 | 5683859 |