Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023258.4 | NP_075747.3 | 3 | 577 | ||
GenBank mRNA | AB032249.1 | - | 3 | 449 | ||
AF310104.1 | - | 3 | 389 | |||
AK007742.1 | - | 3 | 447 | |||
AK009852.1 | - | 3 | 452 | |||
AK018682.1 | - | 3 | 457 | |||
AK152677.1 | - | 3 | 437 | |||
BC008252.1 | - | 3 | 419 | 2648391 |