Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023755.2 | NP_076244.2 | 15 | 4993 | ||
GenBank mRNA | AF311309.1 | - | 15 | 5028 | ||
AK143960.1 | - | 15 | 4989 | |||
BC116663.1 | - | 15 | 4979 | 40087602 |