Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023900.2 | NP_076389.2 | 4, 5 | 780 | ||
GenBank mRNA | AK020621.1 | - | 4, 5 | 449 | ||
AK051734.1 | - | 4, 5 | 437 | |||
AK088567.1 | - | 4, 5 | 743 | |||
AK150144.1 | - | 4, 5 | 395 | |||
AK178993.1 | - | 2, 3 | 185 | |||
AK188371.1 | - | 1 | 16 | |||
AK219000.1 | - | 2, 3 | 117 | |||
BC005565.1 | - | 4, 5 | 378 | 3966305 |