Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard3nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002760.1 | - | 8 | 1018 | ||
AK152116.1 | - | 9 | 1081 | |||
AK152378.1 | - | 9 | 1032 | |||
BC003334.1 | - | 9 | 1035 | 3256725 |