Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005493.2 | NP_005484.2 | 6 | 1117 | ||
XM_006714945.2 | XP_006715008.1 | 6 | 597 | |||
XM_011514205.2 | XP_011512507.1 | 6 | 1135 | |||
XM_017010149.1 | XP_016865638.1 | 6 | 505 | |||
GenBank mRNA | AB008515.1 | - | 6 | 806 | ||
AB055311.1 | - | 6 | 1117 | |||
AF306510.1 | - | 6 | 655 | |||
AK313334.1 | - | 5 | 508 | |||
BC019886.1 | - | 6 | 571 | |||
BC052781.1 | - | 6 | 1278 | |||
BC063849.1 | - | 4 | 373 | 6186252 | ||
KU921435.1 | - | 1 | 131 |