Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190264.1 | NP_001177193.1 | 4 | 425 | ||
NM_025369.3 | NP_079645.1 | 4 | 431 | |||
GenBank mRNA | AK003111.1 | - | 4 | 361 | ||
AK003765.1 | - | 4 | 408 | |||
AK007512.1 | - | 4 | 402 | |||
AK009050.1 | - | 4 | 410 | |||
AK009966.1 | - | 4 | 356 | |||
BC028499.1 | - | 4 | 387 | 1382215 | ||
BC096576.1 | - | 4 | 396 | 1401014 |