Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291197.1 | NP_001278126.1 | 8 | 1238 | ||
NM_001291200.1 | NP_001278129.1 | 7 | 1193 | |||
NM_001291201.1 | NP_001278130.1 | 7 | 1178 | |||
NM_001291202.1 | NP_001278131.1 | 7 | 1175 | |||
NM_001291203.1 | NP_001278132.1 | 6 | 1133 | |||
NM_001291204.1 | NP_001278133.1 | 5 | 984 | |||
NM_025482.3 | NP_079758.2 | 8 | 1235 | |||
GenBank mRNA | AK013085.1 | - | 8 | 1171 | ||
AK013188.1 | - | 8 | 1188 | |||
AK051268.1 | - | 7 | 1119 | |||
AK147885.1 | - | 7 | 1153 | |||
AK155987.1 | - | 6 | 1070 | |||
AK161092.1 | - | 6 | 1089 | |||
BC006886.1 | - | 8 | 1124 | 2647702 |