Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291197.1 | NP_001278126.1 | 8 | 1555 | ||
NM_001291200.1 | NP_001278129.1 | 7 | 1510 | |||
NM_001291201.1 | NP_001278130.1 | 7 | 1495 | |||
NM_001291202.1 | NP_001278131.1 | 7 | 1492 | |||
NM_001291203.1 | NP_001278132.1 | 6 | 1450 | |||
NM_001291204.1 | NP_001278133.1 | 5 | 1301 | |||
NM_025482.3 | NP_079758.2 | 8 | 1552 | |||
GenBank mRNA | AK051268.1 | - | 7 | 1436 | ||
AK147885.1 | - | 7 | 1470 | |||
AK155987.1 | - | 6 | 1387 | |||
AK161092.1 | - | 6 | 1406 | |||
BC006886.1 | - | 8 | 1441 | 2647702 |