Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nosip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163684.1 | NP_001157156.1 | 9 | 948 | ||
NM_025533.3 | NP_079809.1 | 9 | 1023 | |||
GenBank mRNA | AK005700.1 | - | 1 | 63 | ||
AK007962.1 | - | 9 | 897 | |||
AK010006.1 | - | 9 | 959 | |||
AK076736.1 | - | 9 | 948 | |||
BC089029.1 | - | 9 | 949 | 30624825 |