Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042592.2 | NP_001036057.1 | 2, 3 | 547 | ||
NM_025549.3 | NP_079825.2 | 2, 3 | 547 | |||
GenBank mRNA | AK010022.1 | - | 2, 3 | 513 | ||
AK010352.1 | - | 2, 3 | 551 | |||
AK037198.1 | - | 2, 3 | 514 | |||
BC017528.1 | - | 2, 3 | 497 | 4922348 | ||
DQ861996.1 | - | 2, 3 | 351 |