Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asrgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025610.3 | NP_079886.2 | 7 | 1751 | ||
GenBank mRNA | AK030209.1 | - | 7 | 1752 | ||
BC016106.1 | - | 5 | 1503 | 4921265 |