Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025807.3 | NP_080083.2 | 3 | 380 | ||
GenBank mRNA | AK004556.1 | - | 3 | 381 | ||
AK004599.1 | - | 3 | 559 | |||
AK004684.2 | - | 3 | 438 | |||
AK015196.1 | - | 1 | 157 | |||
BC034205.1 | - | 3 | 335 | 4987055 | ||
BC055839.1 | - | 3 | 338 | 4237811 |