Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025807.3 | NP_080083.2 | 5 | 814 | ||
GenBank mRNA | AK004556.1 | - | 5 | 815 | ||
AK004599.1 | - | 5 | 993 | |||
AK004684.2 | - | 5 | 872 | |||
AK015196.1 | - | 3 | 591 | |||
BC034205.1 | - | 5 | 769 | 4987055 | ||
BC055839.1 | - | 5 | 772 | 4237811 |