Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025836.3 | NP_080112.1 | 5 | 607 | ||
GenBank mRNA | AK004970.1 | - | 5 | 608 | ||
AK009148.1 | - | 1 | 154 | |||
AK012517.1 | - | 5 | 608 | |||
AK049537.1 | - | 5 | 608 | |||
AK050140.1 | - | 5 | 609 | |||
AK132313.1 | - | 608 | ||||
AK167214.1 | - | 5 | 606 | |||
AK197135.1 | - | 3 | 365 | |||
AK203053.1 | - | 3 | 345 | |||
BC011116.1 | - | 5 | 603 | 4023354 |