Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025873.2 | NP_080149.2 | 2 | 221 | ||
GenBank mRNA | AK003556.1 | - | 2 | 222 | ||
AK010176.1 | - | 2 | 221 | |||
AK128921.1 | - | 2 | 220 | |||
BC019812.1 | - | 2 | 190 | 5042856 | ||
BC051040.1 | - | 2 | 197 |