Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025920.3 | NP_080196.3 | 3 | 1672 | ||
GenBank mRNA | AK008304.1 | - | 1 | 56 | ||
AK008587.1 | - | 1 | 55 | |||
AK086483.1 | - | 3 | 1486 | |||
AK140888.1 | - | 1615 | ||||
AK167814.1 | - | 3 | 1432 | |||
BC013538.1 | - | 2 | 509 | |||
BC057963.1 | - | 2 | 970 | |||
BC063758.1 | - | 3 | 1403 | 6334411 | ||
BC066042.1 | - | 3 | 1607 | 6413779 |