Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025982.4 | NP_080258.1 | 6 | 1286 | ||
GenBank mRNA | AK012457.1 | - | 6 | 1287 | ||
AK012567.1 | - | 6 | 1289 | |||
AK012853.1 | - | 7 | 1188 | |||
AK014183.1 | - | 6 | 1284 | |||
AK160269.1 | - | 3 | 1975 | |||
BC023306.1 | - | 6 | 1263 | 4165774 |