Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025994.3 | NP_080270.2 | 4 | 2354 | ||
GenBank mRNA | AK048475.1 | - | 4 | 2355 | ||
AK159367.1 | - | 4 | 2344 | |||
AK167267.1 | - | 4 | 2346 |