Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026033.2 | NP_080309.2 | 5 | 1162 | ||
GenBank mRNA | AB047921.1 | - | 5 | 1143 | ||
AK012921.1 | - | 5 | 1163 | |||
AK078416.1 | - | 5 | 1107 |