Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026295.2 | NP_080571.2 | 13 | 3336 | ||
GenBank mRNA | AK016213.1 | - | 4 | 858 | ||
AK017665.1 | - | 2 | 545 | |||
AK080231.1 | - | 1 | 239 | |||
AK156617.1 | - | 13 | 3365 | |||
BC052934.1 | - | 4 | 941 | |||
BC053435.1 | - | 13 | 3336 | 6394578 |