Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026341.2 | NP_080617.2 | 2 | 186 | ||
GenBank mRNA | AK014204.1 | - | 3 | 409 | ||
AK017031.1 | - | 2 | 155 | |||
AK038639.1 | - | 1 | 168 | |||
BC021366.1 | - | 3 | 320 | |||
BC037091.1 | - | 1 | 109 | |||
BC038023.1 | - | 3 | 268 | |||
BC040807.1 | - | 2 | 133 | |||
BC049948.1 | - | 2 | 140 | 3666660 | ||
BC056982.1 | - | 3 | 315 | |||
BF323208.1 | - | 2 | 165 |