Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164139.1 | NP_001157611.1 | 3 | 341 | ||
NM_001164140.1 | NP_001157612.1 | 2 | 260 | |||
NM_001164141.1 | NP_001157613.1 | 3 | 349 | |||
NM_026457.2 | NP_080733.2 | 2 | 199 | |||
GenBank mRNA | AK005703.1 | - | 2 | 198 | ||
AK007022.1 | - | 2 | 178 | |||
AY092416.1 | - | 2 | 333 | |||
BC109147.2 | - | 2 | 178 | 40045644 |