Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026483.2 | NP_080759.2 | 10 | 1731 | ||
GenBank mRNA | AK008785.1 | - | 7 | 661 | ||
AK013337.1 | - | 10 | 1719 | |||
AV222458.2 | - | 3 | 222 | |||
BC018177.1 | - | 10 | 1725 | |||
BC029198.1 | - | 2 | 120 | |||
BC049270.1 | - | 10 | 1707 | 3988645 |