Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026483.2 | NP_080759.2 | 11 | 1953 | ||
GenBank mRNA | AK008785.1 | - | 8 | 883 | ||
AV222458.2 | - | 4 | 444 | |||
BC029198.1 | - | 3 | 342 | |||
BC049270.1 | - | 11 | 1929 | 3988645 |