Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037098.1 | NP_001032175.1 | 6 | 2700 | ||
NM_026495.3 | NP_080771.3 | 6 | 2177 | |||
GenBank mRNA | AK002942.1 | - | 1 | 336 | ||
AK147293.1 | - | 6 | 2177 | |||
AK147310.1 | - | 6 | 2281 | |||
BC022103.1 | - | 6 | 2159 | 5067063 | ||
BC070453.1 | - | 6 | 2480 |