Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026559.3 | NP_080835.1 | 4 | 878 | ||
GenBank mRNA | AK007595.1 | - | 4 | 882 | ||
AK076425.1 | - | 4 | 878 |