Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yeats4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026570.4 | NP_080846.1 | 2, 3 | 362 | ||
GenBank mRNA | AK010522.1 | - | 2, 3 | 363 | ||
AK012096.1 | - | 2, 3 | 329 | |||
AK081867.1 | - | 2, 3 | 345 | |||
AK182917.1 | - | 1, 2 | 15 | |||
AK219066.1 | - | 1, 2 | 15 | |||
BC020043.1 | - | 2, 3 | 330 | 4013548 |