Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026752.4 | NP_081028.3 | 3, 4 | 423 | ||
GenBank mRNA | AK003661.1 | - | 3, 4 | 320 | ||
AK139356.1 | - | 358 | ||||
AK155300.1 | - | 3, 4 | 424 | |||
AK171099.1 | - | 3, 4 | 364 | |||
AK192012.1 | - | 1, 2 | 34 | |||
AK197357.1 | - | 1, 2 | 34 | |||
AK216869.1 | - | 1, 2 | 155 | |||
BC019521.1 | - | 3, 4 | 353 | 4220205 | ||
BC132248.1 | - | 3, 4 | 323 | 40130525 | ||
BC132250.1 | - | 3, 4 | 323 | 40130527 |