Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026752.4 | NP_081028.3 | 7 | 955 | ||
GenBank mRNA | AK003661.1 | - | 7 | 852 | ||
AK139356.1 | - | 947 | ||||
AK155300.1 | - | 7 | 956 | |||
AK171099.1 | - | 7 | 896 | |||
BC019521.1 | - | 7 | 885 | 4220205 | ||
BC132248.1 | - | 7 | 855 | 40130525 | ||
BC132250.1 | - | 7 | 855 | 40130527 |