Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026850.4 | NP_081126.2 | 3 | 239 | ||
GenBank mRNA | AF110512.1 | - | 2 | 209 | ||
AK004334.1 | - | 3 | 234 | |||
AK078372.1 | - | 3 | 240 | |||
AK168509.1 | - | 3 | 280 | |||
BC005601.1 | - | 3 | 260 | 3483084 |